Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NIPSNAP1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501984298
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-4298 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-4298 Supplier Bioss Supplier No. BS19252R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NIPSNAP is a member of an evolutionarily well conserved gene family and has a strong sequence similarity to the central portion of a protein encoded by C. elegans chromosome III between a 4-nitrophenylphosphatase (NIP) domain and non-neuronal SNAP25-like protein. Recent studies have indicated that NIPSNAP is involved in the regulation of the Ca2+-selective transient receptor potential vanilloid channel 6 (TRPV6). NIPSNAP1 associates with TRPV6 at the plasma membrane and inhibits TRPV6 currents. Other studies show that NIPSNAP’s expression is reduced in the phenylketonuria (PKU) mouse brain, suggesting that NIPSNAP may play a role in memory.

Specifications

Antigen NIPSNAP1
Applications Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene NIPSNAP1
Gene Accession No. Q9BPW8
Gene Alias 4-nitrophenylphosphatase domain and non-neuronal SNAP25-like 1; 4-nitrophenylphosphatase domain and non-neuronal SNAP25-like protein homolog 1; 4-nitrophenylphosphatase domain and non-neuronal SNAP25-like protein homolog 1 (C. elegans); nipsnap homolog 1 (C. elegans); Nipsnap1; Protein NipSnap homolog 1
Gene Symbols NIPSNAP1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human NIPSNAP1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8508
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.