Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NIPSNAP3B Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501984701
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-4701 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-4701 Supplier Bioss Supplier No. BS24098R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The NIPSNAP proteins comprise a family of evolutionarily well-conserved proteins with strong sequence similarity to the central portion of a protein encoded by C. elegans chromosome III between a 4-nitrophenylphosphatase (NIP) domain and non-neuronal SNAP25-like protein. NIPSNAP2, a novel gene encoding a protein with tyrosine phosphorylation sites and a transmembrane domain, is co-amplified with EGFR in approximately 40% of glioblastomas, the most common and malignant form of central nervous system tumors. While NIPSNAP3B is highly expressed skeletal muscle, NIPSNAP3A mRNA levels are low. NIPSNAP proteins have been suggested to be important in vesicular transport.

Specifications

Antigen NIPSNAP3B
Applications Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene NIPSNAP3B
Gene Accession No. Q9BS92
Gene Alias 2700063N13Rik; FP944; nipsnap homolog 3A; nipsnap homolog 3B; nipsnap homolog 3B (C. elegans); NIPSNAP3; Nipsnap3a; NIPSNAP3B; NipSnap-related protein; Protein NipSnap homolog 3A; Protein NipSnap homolog 3B; SNAP1
Gene Symbols NIPSNAP3B
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from mouse NIPSNAP3B.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55335
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.