Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NIR1 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS8513R

Description
This gene encodes a member of a family of membrane-associated phosphatidylinositol transfer domain-containing proteins. The calcium-binding protein has phosphatidylinositol transfer activity and interacts with the protein tyrosine kinase PTK2B . The protein is homologous to a Drosophila protein that is implicated in the visual transduction pathway in flies. Mutations in this gene result in autosomal dominant cone dystrophy. Multiple transcript variants encoding different isoforms have been found for this gene.Specifications
NIR1 | |
Polyclonal | |
Unconjugated | |
PITPNM3 | |
ACKR6; atypical chemokine receptor 6; cone rod dystrophy 5; CORD5; Membrane-associated phosphatidylinositol transfer protein 3; MGC157740; MGC157741; NIR1; NIR-1; Phosphatidylinositol transfer protein, membrane-associated 3; PITPnm 3; PITPNM family member 3; PITPNM3; Pyk2 N-terminal domain-interacting receptor 1; RDGBA3; retinal degeneration B alpha 3 | |
Rabbit | |
Protein A | |
RUO | |
83394 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q9BZ71 | |
PITPNM3 | |
KLH conjugated synthetic peptide derived from human NIR1/RDGBA3. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction