Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nir2 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 269831AP150UL
Description
Regulates RHOA activity, and plays a role in cytoskeleton remodeling. Necessary for normal completion of cytokinesis. Plays a role in maintaining normal diacylglycerol levels in the Golgi apparatus. Binds phosphatidyl inositol phosphates (in vitro). May catalyze the transfer of phosphatidylinositol and phosphatidylcholine between membranes. Necessary for maintaining the normal structure of the endoplasmic reticulum and the Golgi apparatus. Required for protein export from the endoplasmic reticulum and the Golgi. Binds calcium ions.Specifications
| Nir2 | |
| Polyclonal | |
| Unconjugated | |
| PITPNM1 | |
| DRES9, NIR 2, NIR2, PITPNM, PITPnm 1, PITPNM1, Rd9, RDGB, RDGB1, RDGBA, RDGBA1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 9600 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.37 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O00562 | |
| PITPNM1 | |
| Nir2 Fusion Protein Ag25357 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction