Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NISCH Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | NISCH |
---|---|
Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:20 - 1:50 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
NISCH Polyclonal specifically detects NISCH in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
NISCH | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human | |
hIRAS, I-1 receptor candidate protein, I-1FLJ14425, I1R, I1R candidate protein, Imidazoline receptor 1, imidazoline receptor antisera selected, Imidazoline receptor antisera-selected protein, imidazoline receptor candidate, Imidazoline-1 receptor, Imidazoline-1 receptor candidate protein, IR1, IRASFLJ40413, KIAA0975FLJ90519, nischarin | |
NISCH | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Immunohistochemistry, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
Polyclonal | |
Rabbit | |
Signal Transduction | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
11188 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:AHFLHFHFYEINGITAALAEELFEKGEQLLGAGEVFAIGPLQLYAVTEQLQQGKPTCASGDAKTDLGHILDFTCRLKYLKVSGTEGPFGTSNIQEQLLPFDLSIFKSLHQVEISHCDAKHIRGLVASKPTLATLSVRFSATSMKEV | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title