Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NISCH Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554718
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554718 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554718 Supplier Invitrogen™ Supplier No. PA554718
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AHFLHFHFYE INGITAALAE ELFEKGEQLL GAGEVFAIGP LQLYAVTEQL QQGKPTCASG DAKTDLGHIL DFTCRLKYLK VSGTEGPFGT SNIQEQLLPF DLSIFKSLHQ VEISHCDAKH IRGLVASKPT LATLSVRFSA TSMKEV Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 93%.

Acts either as the functional imidazoline-1 receptor (I1R) candidate or as a membrane-associated mediator of the I1R signaling. Binds numerous imidazoline ligands that induces initiation of cell-signaling cascades triggering to cell survival, growth and migration. Its activation by the agonist rilmenidine induces an increase in phosphorylation of mitogen-activated protein kinases MAPK1 and MAPK3 in rostral ventrolateral medulla (RVLM) neurons that exhibited rilmenidine-evoked hypotension. Blocking its activation with efaroxan abolished rilmenidine-induced mitogen-activated protein kinase phosphorylation in RVLM neurons. Acts as a modulator of Rac-regulated signal transduction pathways. Suppresses Rac1-stimulated cell migration by interacting with PAK1 and inhibiting its kinase activity. Also blocks Pak-independent Rac signaling by interacting with RAC1 and inhibiting Rac1-stimulated NF-kB response element and cyclin D1 promoter activation. Inhibits also LIMK1 kinase activity by reducing LIMK1 'Tyr-508' phosphorylation. Inhibits Rac-induced cell migration and invasion in breast and colon epithelial cells. Inhibits lamellipodia formation, when overexpressed. Plays a role in protection against apoptosis. Involved in association with IRS4 in the enhancement of insulin activation of MAPK1 and MAPK3. When overexpressed, induces a redistribution of cell surface ITGA5 integrin to intracellular endosomal structures.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NISCH
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NISCH
Gene Accession No. Q9Y2I1
Gene Alias 1200007D05Rik; 3202002H23Rik; AW494485; hIRAS; I-1; I-1 receptor candidate protein; I1R; I1R candidate protein; imidazoline receptor 1; imidazoline receptor antisera selected; Imidazoline receptor antisera-selected protein; imidazoline receptor I-1; imidazoline receptor I-1-like protein; Imidazoline-1 receptor; Imidazoline-1 receptor candidate protein; IR1; IRAS; KIAA0975; NISCH; Nischarin
Gene Symbols NISCH
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NISCH. Recombinant protein control fragment (Product #RP-91062).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11188
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.