Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NISCH Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
| Antigen | NISCH |
|---|---|
| Concentration | 0.13 mg/mL |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-172-9021
![]()
|
Proteintech Group Inc
138131AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Acts either as the functional imidazoline-1 receptor (I1R) candidate or as a membrane-associated mediator of the I1R signaling. Binds numerous imidazoline ligands that induces initiation of cell-signaling cascades triggering to cell survival, growth and migration. Its activation by the agonist rilmenidine induces an increase in phosphorylation of mitogen-activated protein kinases MAPK1 and MAPK3 in rostral ventrolateral medulla (RVLM) neurons that exhibited rilmenidine-evoked hypotension. Blocking its activation with efaroxan abolished rilmenidine-induced mitogen-activated protein kinase phosphorylation in RVLM neurons. Acts as a modulator of Rac-regulated signal transduction pathways. Suppresses Rac1-stimulated cell migration by interacting with PAK1 and inhibiting its kinase activity. Also blocks Pak-independent Rac signaling by interacting with RAC1 and inhibiting Rac1-stimulated NF-kB response element and cyclin D1 promoter activation. Inhibits also LIMK1 kinase activity by reducing LIMK1 ′Tyr-508′ phosphorylation. Inhibits Rac-induced cell migration and invasion in breast and colon epithelial cells. Inhibits lamellipodia formation, when overexpressed. Plays a role in protection against apoptosis. Involved in association with IRS4 in the enhancement of insulin activation of MAPK1 and MAPK3. When overexpressed, induces a redistribution of cell surface ITGA5 integrin to intracellular endosomal structures.Specifications
| NISCH | |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| Q4G017, Q80TM9, Q9Y2I1 | |
| 11188, 306255, 64652 | |
| NISCH Fusion Protein Ag4880 | |
| Primary | |
| -20°C | |
| NISCH |
| 0.13 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| hIRAS, I 1, I 1 receptor candidate protein, I1R, I1R candidate protein, Imidazoline 1 receptor, Imidazoline receptor 1, IR1, IRAS, KIAA0975, NISCH, nischarin | |
| Nisch | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title