Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Reichert™ NIST Certified Calibration Oil

Catalog No. 13940039 Shop All Reichert, Inc Products
Change view
Click to view available options
Refractive Index:
1.35999
1.40424
1.45755
1.48993
1.51432
1.57230
1.63040
Scale Range:
18.04 Brix
42.21 Brix
67.11 Brix
79.70 Brix
89.16 Brix
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Refractive Index Scale Range
13-940-039 1.48993 79.70 Brix
13-940-036 1.51432 89.16 Brix
13-940-038 1.45755 67.11 Brix
13-940-037 1.40424 42.21 Brix
13-940-040 1.57230 18.04 Brix
13-940-041 1.63040 18.04 Brix
13-940-042 1.35999 18.04 Brix
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 13-940-039 Supplier Reichert™ Supplier No. 13K41360
Add to Cart
Add to Cart

Check the accuracy of an instrument using calibration oils. Reichert™ NIST Certified Calibration Oil are formulated laser liquid calibration oils that are supplied with an NIST certification of traceability.

Formulated, measured, and certified to five significant places of refractive index (0.00001).

Specifications

Type NIST Certified Calibration Oil
Scale Range 79.70 Brix
Refractive Index 1.48993
For Use With (Equipment) Refractometer
Includes NIST certification of traceability
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.