Learn More
Beckman Coulter NIST Traceable Particle Size Standards
Recommended for LS Series Instruments to verify instrument and module performance.
Supplier: Beckman Coulter 7800370
Description
- For verifying instrument and module performance
- Recommended for LS Series instruments
- NIST traceable
Specifications
Particle Size Standard, G15 | |
15 μm | |
Verifying Instrument and Module Performance |
Silicon Carbide | |
10 Pk. | |
LS Series Instruments with ULM |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.