Learn More
Beckman Coulter NIST Traceable Particle Size Standards
Catalog No. p-7215335
Click to view available options
Particle Size:
15 μm
35 μm
300 μm
500 μm
Material:
Glass
Polystyrene
Silicon Carbide
This item is not returnable. View return policy
This item is not returnable. View return policy
Recommended for LS Series Instruments to verify instrument and module performance.
  • For verifying instrument and module performance
  • Recommended for LS Series instruments
  • NIST traceable
TRUSTED_SUSTAINABILITY
Specifications
Product Type Particle Size Standard, GB500
Material Glass
Particle Size 500 μm
Quantity 5 Pk.
For Use With (Application) Verifying Instrument and Module Performance
For Use With (Equipment) LS Series Instruments with ULM & DPS
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.