Learn More
Beckman Coulter NIST Traceable Particle Size Standards
Catalog No. 15655215
Change view
Click to view available options
Particle Size:
15 μm
35 μm
300 μm
500 μm
Material:
Glass
Polystyrene
Silicon Carbide
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Particle Size Material
15-655-215 300 μm Polystyrene
15-655-216 500 μm Glass
15-655-214 15 μm Silicon Carbide
15-655-217 35 μm Silicon Carbide
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 15-655-215 Supplier Beckman Coulter Supplier No. 7800377
This item is not returnable. View return policy
Recommended for LS Series Instruments to verify instrument and module performance.
  • For verifying instrument and module performance
  • Recommended for LS Series instruments
  • NIST traceable
Specifications
Product Type Particle Size Standard, Latron 300
Material Polystyrene
Particle Size 300 μm
Quantity 12 Pk.
For Use With (Application) Verifying Instrument and Module Performance
For Use With (Equipment) LS Series Instruments with ULM