Learn More
Beckman Coulter NIST Traceable Particle Size Standards
Catalog No. 15655217
Change view
Click to view available options
Particle Size:
15 μm
35 μm
300 μm
500 μm
Material:
Glass
Polystyrene
Silicon Carbide
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Particle Size Material
15-655-217 35 μm Silicon Carbide
15-655-216 500 μm Glass
15-655-215 300 μm Polystyrene
15-655-214 15 μm Silicon Carbide
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 15-655-217 Supplier Beckman Coulter Supplier No. 7800499/DEL
Recommended for LS Series Instruments to verify instrument and module performance.
  • For verifying instrument and module performance
  • Recommended for LS Series instruments
  • NIST traceable
TRUSTED_SUSTAINABILITY
Specifications
Product Type Particle Size Standard, G35
Material Silicon Carbide
Particle Size 35 μm
Quantity 5 Pk.
For Use With (Application) Verifying Instrument and Module Performance
For Use With (Equipment) LS Series Instruments with DPS