Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NIT1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552317
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552317 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552317 Supplier Invitrogen™ Supplier No. PA552317
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FLSLLCPGLR IPQLSVLCAQ PRPRAMAISS SSCELPLVAV CQVTSTPDKQ QNFKTCAELV REAARLGACL AFLPEAFDFI ARDPAETLHL SEPLGGKLLE EYTQLARECG LWLSLGGFHE RGQDWEQT Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 67%.

Plays a role in cell growth and apoptosis: loss of expression promotes cell growth and resistance to DNA damage stress. Has tumor suppressor properties that enhances the apoptotic responsiveness in cancer cells; this effect is additive to the tumor suppressor activity of FHIT. It is also a negative regulator of primary T-cells. Has apparently no omega-amidase activity such as NIT2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NIT1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NIT1
Gene Accession No. Q86X76
Gene Alias AI255805; Deaminated glutathione amidase; dGSH amidase; ESTM30; MGC57670; Nit1; nitrilase 1; nitrilase homolog 1; rCG_20134; W57327
Gene Symbols NIT1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NIT1. Recombinant protein control fragment (Product #RP-89178).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4817
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.