Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NIT2 Monoclonal Antibody (OTI2B9), TrueMAB™, OriGene
SDP

Catalog No. 502542531
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-2531 30 μL
50-167-2252 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-2531 Supplier OriGene Supplier No. TA501109S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NIT2 Monoclonal antibody specifically detects NIT2 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen NIT2
Applications Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI2B9
Concentration 0.74 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene PRKAR2A
Gene Accession No. P13861
Gene Alias 1190017B19Rik; CUA002; D16Ertd502e; epididymis secretory sperm binding protein Li 8a; HEL-S-8a; Nit protein 2; NIT2; nitrilase family member 2; nitrilase family, member 2; nitrilase homolog 2; Omega-amidase NIT2; RGD1310494
Host Species Mouse
Immunogen Full length human recombinant protein of human PRKAR2A produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5576
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.