Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NIT2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA549733
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA549733 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA549733 Supplier Invitrogen™ Supplier No. PA549733
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody detects endogenous level of total NIT2 protein.

Has a omega-amidase activity. The role of omega-amidase is to remove potentially toxic intermediates by converting alpha-ketoglutaramate and alpha-ketosuccinamate to biologically useful alpha-ketoglutarate and oxaloacetate, respectively. Overexpression decreases the colony-forming capacity of cultured cells by arresting cells in the G2 phase of the cell cycle.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NIT2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene Nit2
Gene Accession No. Q9JHW2, Q9NQR4
Gene Alias 1190017B19Rik; CUA002; D16Ertd502e; epididymis secretory sperm binding protein Li 8a; HEL-S-8a; Nit protein 2; NIT2; nitrilase family member 2; nitrilase family, member 2; nitrilase homolog 2; Omega-amidase NIT2; RGD1310494
Gene Symbols Nit2
Host Species Rabbit
Immunogen Synthetic peptide corresponding to a region derived from 264-276 amino acids of Human itrilase family, member 2.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 52633, 56954
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.