Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NIT2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA557908
Description
Immunogen sequence: YLIGGSIPEE DAGKLYNTCA VFGPDGTLLA KYRKIHLFDI DVPGKITFQE SKTLSPGDSF STFDTPYCRV GLGICYDMRF AELAQIYAQ Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 94%.
Has a omega-amidase activity. The role of omega-amidase is to remove potentially toxic intermediates by converting alpha-ketoglutaramate and alpha-ketosuccinamate to biologically useful alpha-ketoglutarate and oxaloacetate, respectively. Overexpression decreases the colony-forming capacity of cultured cells by arresting cells in the G2 phase of the cell cycle.
Specifications
NIT2 | |
Polyclonal | |
Unconjugated | |
Nit2 | |
1190017B19Rik; CUA002; D16Ertd502e; epididymis secretory sperm binding protein Li 8a; HEL-S-8a; Nit protein 2; NIT2; nitrilase family member 2; nitrilase family, member 2; nitrilase homolog 2; Omega-amidase NIT2; RGD1310494 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
288174, 52633, 56954 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q497B0, Q9JHW2, Q9NQR4 | |
Nit2 | |
Recombinant protein corresponding to Human NIT2. Recombinant protein control fragment (Product #RP-96272). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction