Learn More
Nitinol foil, 0.05mm (0.002in) thick, superelastic, flat annealed, pickled surface
Description
Nitinol foil is used in cell-phone technology as a retractable antenna, or microphone boom, due to its highly flexible and mechanical memory nature. It is also used in some novelty products, such as self-bending spoons which can be used by amateur and stage magicians to demonstrate psychic powers or as a practical joke, as the spoon will bend itself when used to stir tea, coffee, or any other warm liquid. It is highly biocompatible and has properties suitable for use in orthopaedic implants. Due to Nitinol's unique properties it has seen a large demand for use in less invasive medical devices.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Chemical Name or Material | Nitinol foil |
Name Note | 0.05mm (0.002 in.) thick, superelastic, flat annealed, pickled surface |
Quantity | 100 x 100 mm |
Odor | Odorless |
Weight | ≈0.25g/25x25mm |
MDL Number | MFCD02091734 |
Solubility Information | Insoluble in water. |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.