Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitinol tension coil spring, wire diameter 0.75mm (0.030in), mean coil diameter 5.55mm (0.22in), Alloy M

Catalog No. AA45655KJ
Click to view available options
Quantity:
100 Pc.
25 Pc.
5 Pc.

CAS: 7440-02-0

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Color Gray
Chemical Name or Material Nitinol tension coil spring
Name Note Wire diameter 0.75mm, mean coil diameter 5.55mm
CAS 7440-02-0, 7440-32-6
Quantity 5 Pc.
Odor Odorless
Form Solid
MDL Number MFCD02091734
Concentration or Composition (by Analyte or Components) Nickel: 55.75%; Titanium: 44.25%

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.