Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitinol tubing, 0.584mm (0.023in) OD, 0.33mm (0.013in) ID, superelastic, Alloy S, straight annealed, oxide surface, Thermo Scientific™

Catalog No. p-7052273
Click to view available options
Quantity:
50 cm
100 cm

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Nitinol tubing
Name Note 0.584mm (0.023 in.) OD, 0.33mm (0.013 in.) ID, superelastic, Alloy S, straight annealed, oxide surface
Quantity 50 cm
Odor Odorless
MDL Number MFCD02091734
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.