Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitinol wire, 1mm, Thermo Scientific™

Catalog No. p-4422429
Change view
Click to view available options
Quantity:
10 m
150 m
50 m
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA45087G6 10 m
AA45087H2 50 m
AA45087H5 150 m
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA45087G6 Supplier Thermo Scientific Chemicals Supplier No. 45087G6

Form: approx. 49.71g/10m; Af approx. 10°C. Solid in various forms

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Nitinol
Color Silver to Gray
Appearance Silver gray wire
Quantity 10 m
Odor Odorless
Form Wire
MDL Number MFCD02091734
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.