Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LaMotte™ Nitrate-Nitrogen Tablet Test Kit Reagent Refill
SDPSCI_ED

Catalog No. s65411
Click to view available options
Range:
1 to 15 ppm

Includes

Reagent Refill Tabs only.

The Nitrate-Nitrogen Tablet Test Kit Reagent Refill for use in colorimetric test systems.

  • Range: 0 to 15 ppm
  • Sensitivity: 1.0 ppm
  • Number of Tests: 50
  • Reagent Refills for use with Nitrate-Nitrogen Kit (3354-01)

Specifications

Includes Reagent Refills for use with Nitrate-Nitrogen Kit (3354-01)
Range 1 to 15 ppm
Test Method Zinc Reduction
Type Reagent Refills
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.