Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitrazine Yellow

Catalog No. p-7041791
Click to view available options
Quantity:
1 g
5 g
25 g

pH indicator | CAS: 5423-07-4 | C16H8N4Na2O11S2 | 542.36 g/mol

Nitrazine Yellow is used to test vaginal pH during late pregnancy to determine the breakage of the amniotic sac. To perform a fecal pH test for diagnosing intestinal infections or other digestive problems. In civil engineering, to determine the carbonatation spread in concrete structures and therefore assess the state of the rebar's passivation film.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Nitrazine Yellow is used to test vaginal pH during late pregnancy to determine the breakage of the amniotic sac. To perform a fecal pH test for diagnosing intestinal infections or other digestive problems. In civil engineering, to determine the carbonatation spread in concrete structures and therefore assess the state of the rebar′s passivation film.

Solubility
Dissolve in water at 1mg/ml

Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5423-07-4
MDL Number MFCD00003938
Synonym C.I. 14890; Delta Yellow

Specifications

Molecular Formula C16H8N4Na2O11S2
Solubility Information Dissolve in water at 1mg/ml
InChI Key XBLIFEQTVVSTIM-QIKYXUGXSA-L
SMILES C1=CC(=C(C=C1[N+](=O)[O-])[N+](=O)[O-])NN=C2C(=CC3=C(C2=O)C=CC(=C3)S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+]
IUPAC Name disodium;3-[(2,4-dinitrophenyl)hydrazinylidene]-4-oxonaphthalene-2,7-disulfonate
Molecular Weight (g/mol) 542.36
PubChem CID 44134855
Formula Weight 542.36
Color Brown to Orange
Quantity 1 g
Physical Form Powder
Chemical Name or Material Nitrazine Yellow
Show More Show Less
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.