Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCR Safety NitriShield™ 6010 Nitrile Disposable Gloves

NitriShield™ medical grade nitrile gloves are used in applications that require resistance to punctures, tears or exposure to certain types of chemicals. Available in green or orange.

Specifications

Applications Healthcare, Laboratory
Certifications/Compliance Class 1 Medical Device
Thickness (English) 4 mil
Thickness (Metric) 0.10 mm
Exterior Finish Partially Textured
View More Specs

Products 6
Catalog Number Mfr. No. Size Color Price Quantity & Availability  
Catalog Number Mfr. No. Size Color Price Quantity & Availability  
19-812-738
View Documents
MCR Safety
6010GM
Medium Green
Case of 1000 for $376.26
N/A
Due to product restrictions, please to purchase or view availability for this product.
19-812-737
View Documents
MCR Safety
6010GL
Large Green
Case of 1000 for $376.26
N/A
Due to product restrictions, please to purchase or view availability for this product.
19-812-740
View Documents
MCR Safety
6010OL
Large Orange
Pack of 100 for $37.67
Case of 10 PK for $376.70
N/A
Due to product restrictions, please to purchase or view availability for this product.
19-812-739
View Documents
MCR Safety
6010GXL
X-Large Green This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.Due to product restrictions, please to purchase or view availability for this product. N/A
19-812-742
View Documents
MCR Safety
6010OS
Small Orange This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.Due to product restrictions, please to purchase or view availability for this product. N/A
19-812-744
View Documents
MCR Safety
6010OXXL
2X-Large Orange This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.Due to product restrictions, please to purchase or view availability for this product. N/A
Description

Description

  • Green or orange nitrile
  • Medical Grade, Class 1
  • Powder free
  • Rolled cuff
  • Latex-free
  • Resistant to punctures and tears
  • Textured grip
Specifications

Specifications

Healthcare, Laboratory
4 mil
Partially Textured
Class 1 Medical Device
0.10 mm
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit