Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitric Acid, 0.6 Normal, Unstandardized, Reagents

Catalog No. 01334603
Change view
Click to view available options
:
Quantity: 500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01-334-603 Quantity: 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-334-603 Supplier Reagents Holdings Llc Supplier No. CN206600500A
Only null left
Add to Cart
Add to Cart

Nitric Acid, 0.6 Normal, Unstandardized, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Nitric Acid, 0.6 Normal, Unstandardized
pH <1
Color Colorless
CAS 7732-18-5, 7697-37-2
Boiling Point 100°C
Packaging Poly Bottle
Specific Gravity 1.0204
Quantity 500 mL
Molecular Formula HNO3/H2O
Physical Form Liquid
Synonym Azotic Acid, Hydrogen nitrate
Grade Reagent
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.