Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nitric Acid, 20% (v/v), Ricca Chemical
Shop All Ricca Chemical Company Products

Click to view available options
Quantity:
500 mL
1 L
2.5 L
Packaging:
Amber Glass Bottle
Safety-coated Glass Bottle
Specifications
Specifications
Color | Colorless to Yellow |
Appearance of Solution | Colorless to Light Yellow Liquid |
Concentration | 20% (v/v) |
CAS Min % | 79.0, 19.0 |
CAS Max % | 81.0, 21.0 |
Packaging | Amber Glass Bottle |
Solvent or Matrix | Aqueous Solution |
Quantity | 1 L |
Physical Form | Liquid |
Synonym | hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide |
Show More |
Safety and Handling
ShelfLife | 24 months |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction