Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitric Acid, 4% v/v in Denatured Ethanol, Reagents

Catalog No. 01334641
Change view
Click to view available options
:
Quantity: 500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01-334-641 Quantity: 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-334-641 Supplier Reagents Holdings Llc Supplier No. CN105600500C
Only null left
Add to Cart
Add to Cart

Nitric Acid, 4% v/v in Denatured Ethanol, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 64-17-5 , 7697-37-2 , 67-63-0
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.07
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
Synonym Nital etchant
IUPAC Name ethanol
SMILES CCO

Specifications

pH <1
Color Colorless
Boiling Point 82°C
Packaging Glass Bottle
Specific Gravity 0.83
Quantity 500 mL
Physical Form Liquid
Percent Purity 4%
Grade Reagent
Chemical Name or Material Nitric Acid, 4% v/v in Denatured Ethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.