Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitric Acid, 5% (v/v) in Denatured Ethanol, Ricca Chemical
SDP

Catalog No. p-3340629 Shop All Ricca Chemical Company Products
Click to view available options
Quantity:
1 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle

Chemical Identifiers

CAS 64-17-5, 7697-37-2, 67-56-1, 7732-18-5
Molecular Formula HNO3
Molecular Weight (g/mol) 63.01
MDL Number MFCD00011349
InChI Key GRYLNZFGIOXLOG-UHFFFAOYSA-N
Synonym hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
PubChem CID 944
ChEBI CHEBI:48107
IUPAC Name nitric acid
SMILES O[N+]([O-])=O

Specifications

Color Colorless to Yellow
CAS Min % 4.0, 90.0, 4.0
CAS Max % 6.0, 91.0, 6.0
Packaging Natural Poly Bottle
Quantity 1 L
Physical Form Liquid
Percent Purity 5% V/V
Grade Laboratory
Concentration or Composition (by Analyte or Components) 5% (v/v) Alcoholic Solution
Chemical Name or Material Nitric Acid
ShelfLife 12 months
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.