Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nitric Acid, 69.0 to 70.0%, BAKER INSTRA-ANALYZED™ Reagent, J.T. Baker™
Click to view available options
Quantity:
500 mL
2.5 L
Packaging:
Glass Bottle
PVC Coated Glass Bottle
Description
- For Trace Metal Analysis
Specifications
Specifications
Packaging | Glass Bottle |
Specific Gravity | 1L = 1.41kg |
Quantity | 500 mL |
Formula Weight | 63.01 |
Percent Purity | 69.0 to 70.0% |
Grade | BAKER INSTRA-ANALYZED™ Reagent |
Chemical Name or Material | Nitric Acid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction