Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitric Acid, 69.0 to 70.0%, BAKER INSTRA-ANALYZED™ Reagent, J.T. Baker™

Catalog No. 02003465 Shop All Avantor Products
Click to view available options
Quantity:
500 mL
2.5 L
Packaging:
Glass Bottle
PVC Coated Glass Bottle

Acids and standards that are purified and characterized for use in trace element analysis in the low parts-per-billion (ppb) range

    For Trace Metal Analysis

Chemical Identifiers

CAS 7697-37-2
Molecular Formula HNO3
Molecular Weight (g/mol) 63.01
MDL Number MFCD00011349
InChI Key GRYLNZFGIOXLOG-UHFFFAOYSA-N
Synonym hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
PubChem CID 944
ChEBI CHEBI:48107
IUPAC Name nitric acid
SMILES O[N+]([O-])=O

Specifications

Packaging Glass Bottle
Specific Gravity 1L = 1.41kg
Quantity 2.5 L
Formula Weight 63.01
Percent Purity 69.0 to 70.0%
Grade BAKER INSTRA-ANALYZED™ Reagent
Chemical Name or Material Nitric Acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.