Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nitric Acid, BAKER INSTRA-ANALYZED™ Plus, J.T. Baker™
Shop All Avantor Products
Click to view available options
Quantity:
500 mL
2.5 L
Packaging:
PFA Bottle
Poly Bottle
Description
- For use in ICP (Inductively Coupled Plasma Spectrometry), Trace Metal Analysis; For Laboratory, Research, or Manufacturing Use
Specifications
Specifications
Packaging | Poly Bottle |
Specific Gravity | 1L = 1.41kg |
Quantity | 2.5 L |
Formula Weight | 63.01 |
Grade | BAKER INSTRA-ANALYZED™ Plus |
Chemical Name or Material | Nitric Acid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction