Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitric Acid Concentrate (Certified) ACS, Fisher Chemical

Catalog No. sa95 Shop All Fisherbrand Products
Click to view available options
Quantity:
100 mL
Packaging:
Glass Bottle
  • A volumetric standard solution, titrated at 25°C to pH 8.2 (phenolphthalein endpoint) against sodium hydroxide, which has been standardized against NIST™ potassium acid phthalate.
  • Each bottle contains exact amount of 1N concentrate required to make1 liter of N/10 (0.1N) solution.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7697-37-2, 7732-18-5
Molecular Formula HNO3
Molecular Weight (g/mol) 63.01
MDL Number MFCD00011349
InChI Key GRYLNZFGIOXLOG-UHFFFAOYSA-N
Synonym hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
PubChem CID 944
ChEBI CHEBI:48107
IUPAC Name nitric acid
SMILES O[N+]([O-])=O

Specifications

Melting Point >0°C
Boiling Point 100°C
Concentration 0.0995 to 0.1005N
Identification Pass Test
Packaging Glass Bottle
Traceability to NIST LOT 84L Potassium acid phthalate
Quantity 100 mL
Physical Form Liquid
UN Number 2031
Grade Certified
Chemical Name or Material Nitric Acid Concentrate, 1N
Show More Show Less
DOTInformation Class 8: Corrosive
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.