Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitric Acid, Fuming (Certified ACS), Fisher Chemical™

Catalog No. a202400 Shop All Fisherbrand Products
Click to view available options
Quantity:
400 mL
2.5 L
Packaging:
Glass Bottle

Catalog No. A202400

Item Discontinued Due to product restrictions, please to purchase or view availability for this product.This item has been discontinued by the supplier.

Chemical Identifiers

CAS 7697-37-2, 7732-18-5, 10102-44-0
Molecular Formula HNO3
Molecular Weight (g/mol) 63.01
MDL Number MFCD00011349
InChI Key GRYLNZFGIOXLOG-UHFFFAOYSA-N
Synonym hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
PubChem CID 944
ChEBI CHEBI:48107
IUPAC Name nitric acid
SMILES O[N+]([O-])=O

Specifications

pH 1.0
Color Colorless Yellow
Melting Point -41°C
Boiling Point 120.5°C
Density 1.41g/cm³
Heavy Metals (as Pb) 5ppm max.
Identification Pass Test
Packaging Glass Bottle
Assay Percent Range ≥90.0%
Quantity 400 mL
Physical Form Liquid
UN Number 2032
Formula Weight 63.01
Percent Purity ≥90.0%
Grade Certified ACS
Chemical Name or Material Nitric Acid
Show More Show Less
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Oxidizer: Contact with combustible/organic material may cause fire. May cause pulmonary edema. Hygroscopic. Containers should be vented periodically in order to overcome pressure buildup. Use personal protective equipment. Keep away from clothing and other combustible materials. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. Move to fresh air. If breathing is difficult, give oxygen.

NFPA

Health:4
Flammability:0
Instability:0
DOTInformation DOT Class 8, : Corrosive
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.