Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitric Acid, Fuming, Certified ACS, Thermo Scientific™

Catalog No. AC611224000
Click to view available options
:
400mL

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 7697-37-2
Density 1.41g/mL
Heavy Metals (as Pb) 5ppm max.
Identification Pass Test
Residue after Ignition 0.002% max.
Vapor Pressure 0.37 to 0.40kPa
Quantity 400mL
Molecular Formula HNO3
Linear Formula HNO3
MDL Number MFCD00011349
Fieser 01,733; 03,212; 04,356; 05,474
Merck Index 15, 6663
Synonym hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
InChI Key GRYLNZFGIOXLOG-UHFFFAOYSA-N
SMILES [N+](=O)(O)[O-]
IUPAC Name nitric acid
Molecular Weight (g/mol) 63.012
PubChem CID 944
ChEBI CHEBI:48107
Formula Weight 63.01
Percent Purity ≥90.0%
Grade ACS Reagent
Chemical Name or Material Nitric acid
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
May intensify fire; oxidizer.
Precautionary Statement GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.