Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nitric acid, TraceSELECT™, for trace analysis, ≥69.0%, Honeywell Fluka™
Click to view available options
Quantity:
500 mL
1 L
2.5 L
6 x 500 mL
Description
TraceSELECT™ Ultra and TraceSELECT
- ultra-pure acids, bases, and salts for smelting and wet digestion in environmental, water, and food analysis
- sample preparation for trace analysis requires reagents of the highest purity; our TraceSELECT Ultra products for ultra-trace analysis at ppb and even ppt levels are produced by sub-boiling distillation
- the acids, bases, and salts in the TraceSELECT series have been developed for sample preparation and analysis in the ppb (μg/kg) trace range
- packaged in high-quality containers appropriate for each product
- the reagents are produced and bottled under clean-room conditions

Specifications
Specifications
Density | 1.41g/mL at 20°C |
Vapor Pressure | 8 mmHg (20°C) |
Quantity | 500 mL |
UN Number | UN2031 |
Formula Weight | 63.01g/mol |
Percent Purity | ≥69% (T) |
Grade | TraceSELECT™ |
Chemical Name or Material | Nitric acid |
Safety and Handling
Hazard Statement | P210-P220-P260-P280-P305 + P351 + P338-P370 + P378 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction