Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nitrilase 1 Antibody (OTI3A11), PE/Cy7, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272999PECY7
Description
Nitrilase 1 Monoclonal specifically detects Nitrilase 1 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.Specifications
Nitrilase 1 | |
Monoclonal | |
PE-Cyanine7 | |
PBS with 0.05% Sodium Azide | |
NIT1 | |
Full length recombinant protein of human Nitrilase 1 (NP_005591) produced in HEK293T cells. | |
0.1 mL | |
4817.0 | |
Store at 4°C in the dark. Do not freeze. |
Flow Cytometry | |
OTI3A11 | |
Flow Cytometry | |
EC 3.5, MGC57670, nitrilase 1, nitrilase homolog 1 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction