Learn More
PIP™ G-Tek™ Aramid Fiber Gloves, Nitrile-Coated MicroSurface Grip
Catalog No. 19103601 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Glove Cuff Color:
Black
Blue
Brown
Red
Yellow
Size:
Small
Medium
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Glove Cuff Color Size
19-103-601 Red Small
19-103-602 Yellow Medium
19-103-603 Brown Large
19-103-604 Black X-Large
19-103-605 Blue 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-103-601 Supplier PIP™ Supplier No. 09K1660/S
Add to Cart
Add to Cart
This item is not returnable. View return policy
Microsurface coated palm and fingertips provide an exceptional grip in dry, wet and oily conditions by reacting like tiny suction cups that attach firmly to the material being handled.
  • Used for material/glass handling, fishing, sanitation, general maintenance, woodworking, waste handling and recycling
  • Medium weight
  • Microsurface coated palm and fingertips
  • Seamless knit construction
  • Cut-resistant
  • Aramid fibers fiber provides high tensile strength
  • Porous nitrile foam coating provides secure grip in oily applications
Specifications
ANSI Abrasion Rating ANSI 4
Certifications/Compliance (Europe) CE
Construction Seamless Knit
Coating Material Nitrile
Color Black, Yellow
Cut Resistance Rating ANSI A3
Hand Specific
Latex Free Yes
Material Aramid Fiber, Nitrile
Product Type Cut-resistant Gloves
Size Small
EN388 Standard EN 388:4343
For Use With (Application) Material handling, glass handling operations, fishing industries, sanitation, general maintenance, Woodworking, waste handling and recycling
Glove Cuff Color Red
Model 09-K1660/S
Puncture Rating ANSI/ISEA 105-16 Level 3
Series G-Tek™ 09-1660