Learn More
Nitrile Examination Disposable Gloves
Shop All Fisherbrand Products
Click to view available options
Size:
Medium
Large
X-Large
Description
- 3.5 gm nitrile gloves
- Ambidextrous with beaded cuff
- Color: blue
- Certifications: 510K, CE Certification, ISO9001 & ISO13485
- Impermeability as per EN455-1: AQL 1.5
Specifications
Specifications
AQL | 1.5 |
Color | Blue |
Disposable | Single-use |
Material | Nitrile |
Size | Large |
FDA 510 (k) | Yes |