Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ North™ Nitrile Isolator Sleeve

Catalog No. 19163026
This item is not returnable. View return policy
Catalog No. 19-163-026 Supplier Honeywell™ Supplier No. NIT2710
This item is not returnable. View return policy

For use with North NitriGuard and Chemsoft Industrial nitrile gloves

Gives the user the flexibility to replace gloves without having to replace sleeves

  • Can be used with different styles and thicknesses of nitrile glove
  • Proprietary North nitrile formulation offers a great combination of chemical resistance, mechanical protection and wearer comfort
  • Provides good protection from a broad range of chernicals, including acids, bases, alcohols, esters and solvents
  • Good abrasion, puncture, cut and snag resistance
  • Compatible with alcohol cleaning practices. Will not degrade or becorne slippery
  • Inherently anti-static - surface resistance is 1011 ohms/sq providing safer working where electro-static build-up is a concern
TRUSTED_SUSTAINABILITY

Specifications

Product Type Isolator Sleeve
Material Nitrile
Color Green
Finish Rough
Thickness 23 mil
For Use With (Application) Food processing or in environments where electro-static build-up is a concern
Certifications/Compliance USDA and FDA regulations, 21 CFR, for use in food processing.
Length (English) 27 in.
Width (English) 10 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.