Learn More
Nitrilotriacetic acid, 99%
Description
It is employed as an excellent chelating agent. It is also used as a complexing agent, metal masking agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
It is employed as an excellent chelating agent. It is also used as a complexing agent, metal masking agent.
Solubility
Slightly soluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed. Store in cool, dry conditions in well ventilated place.
Specifications
Specifications
| Melting Point | 246°C (decomposition) |
| Odor | Odorless |
| Linear Formula | N(CH2CO2H)3 |
| Quantity | 2500 g |
| Beilstein | 1710776 |
| Merck Index | 14,6579 |
| Solubility Information | Slightly soluble in water. |
| Formula Weight | 191.14 |
| Percent Purity | 99% |
| Chemical Name or Material | Nitrilotriacetic acid |
Safety and Handling
| Hazard Category | H319-H350-H500 |
| Hazard Statement | GHS H Statement H351-H302-H315-H319-H335 Suspected of causing cancer. Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P201-P202-P264b-P280i-P281-P305+P351+P338-P308+P313-P501c |
| EINECSNumber | 205-355-7 |
| RTECSNumber | AJ0175000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.