Alfa Aesar™ Nitrilotriacetic acid, 99%

Manufacturer: Alfa Aesar™ A1193622

Quantity 100g

 View more versions of this product

Catalog No. AAA1193622

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 139-13-9
    Molecular Formula C6H9NO6
    Linear Formula N(CH2CO2H)3
    Formula Weight 191.14
    MDL Number MFCD00004287
    Beilstein 1710776
    Merck Index 14,6579
    Synonym Tris(carboxymethyl)amine; N,N-Bis(carboxymethyl)glycine
    Chemical Name or Material Nitrilotriacetic acid

    Specifications

    Formula Weight 191.14
    Health Hazard 3 GHS P Statement
    P261-P280-P281-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Use personal protective equipment as required.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    RTECS Number AJ0175000
    Beilstein 1710776
    MDL Number MFCD00004287
    CAS 139-13-9
    Synonym Tris(carboxymethyl)amine; N,N-Bis(carboxymethyl)glycine
    Health Hazard 2 GHS H Statement
    H351-H302-H315-H319-H335
    Suspected of causing cancer.
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Quantity 100g
    TSCA TSCA
    Molecular Formula C6H9NO6
    Merck Index 14,6579
    Health Hazard 1 Warning
    Melting Point 246°C (dec.)
    Linear Formula N(CH2CO2H)3
    Odor Odorless
    Chemical Name or Material Nitrilotriacetic acid
    Assay 99%

    Description

    Applications
    It is employed as an excellent chelating agent. It is also used as a complexing agent, metal masking agent.

    Solubility
    Slightly soluble in water.

    Notes
    Store away from oxidizing agents. Keep the container tightly closed. Store in cool, dry conditions in well ventilated place.

    GHS H Statement
    H351-H302-H315-H319-H335
    Suspected of causing cancer.
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P281-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Use personal protective equipment as required.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit