Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitrilotriacetic acid, 99%

Catalog No. p-7081795
Click to view available options
Quantity:
100 g
500 g
2500 g

CAS: 139-13-9 | C6H9NO6 | 191.14 g/mol

It is employed as an excellent chelating agent. It is also used as a complexing agent, metal masking agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as an excellent chelating agent. It is also used as a complexing agent, metal masking agent.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed. Store in cool, dry conditions in well ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 139-13-9
Molecular Formula C6H9NO6
Molecular Weight (g/mol) 191.14
MDL Number MFCD00004287
InChI Key MGFYIUFZLHCRTH-UHFFFAOYSA-N
Synonym nitrilotriacetic acid, 2,2',2-nitrilotriacetic acid, triglycollamic acid, aminotriacetic acid, complexon i, trilon a, n,n-bis carboxymethyl glycine, nitrilotriacetate, komplexon i, titriplex i
PubChem CID 8758
ChEBI CHEBI:44557
IUPAC Name 2-[bis(carboxymethyl)amino]acetic acid
SMILES OC(=O)CN(CC(O)=O)CC(O)=O

Specifications

Melting Point 246°C (decomposition)
Odor Odorless
Linear Formula N(CH2CO2H)3
Quantity 100 g
Beilstein 1710776
Merck Index 14,6579
Solubility Information Slightly soluble in water.
Formula Weight 191.14
Percent Purity 99%
Chemical Name or Material Nitrilotriacetic acid
Hazard Category H319-H350-H500
Hazard Statement GHS H Statement
H351-H302-H315-H319-H335
Suspected of causing cancer.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P201-P202-P264b-P280i-P281-P305+P351+P338-P308+P313-P501c
EINECSNumber 205-355-7
RTECSNumber AJ0175000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.