Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitrilotriacetic acid, trisodium salt, monohydrate, 99+%

Catalog No. p-4510067
Click to view available options
Quantity:
25 g
500 g
2.5 kg
Packaging:
Plastic bottle

Nitrilotriacetic acid, trisodium salt, monohydrate, CAS # 18662-53-8, is considered a non-genotoxic compound showing carcinogenic activity and is used as an inducer of tumorigenesis in laboratory studies using animal models. | CAS: 18662-53-8 | C6H6NNa3O6·H2O | 275.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Information

• Nitrilotriacetic acid, trisodium salt, monohydrate is considered to be a nongenotoxic compound with carcinogenic activity
• This compound inhibits gap-junctional communication between Chinese hamster lung fibroblasts

Applications

• Nitrilotriacetic acid trisodium salt monohydrate is commonly used in laboratory experiments due to its capacity to induce the tumorigenic process
• It has been used in in vivo experiments as an inducer of bladder and kidney tumors in animal models
• It is used as a chelating agent, which forms coordination compounds with metal ions (chelates)

Literature References
Malcolm, A.; Mills, L. Inhibition of gap-junctional intercellular communication between Chinese hamster lung fibroblasts by di(2-ethylhexyl) phthalate (DEHP) and trisodium nitrilotriacetate monohydrate (NTA). Cell Biology and Toxicology. 1989, 5, 145–153.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18662-53-8
Molecular Formula C6H6NNa3O6·H2O
Molecular Weight (g/mol) 275.09
InChI Key HERBOKBJKVUALN-UHFFFAOYSA-K
Synonym trisodium nitrilotriacetate monohydrate, nitrilotriacetic acid trisodium salt monohydrate, nta sodium hydrate, nta trisodium salt dihydrate, unii-j0u92u6nu5, ccris 437, nitrilotriacetic acid trisodium salt, monohydrate, nitriloacetic acid trisodium salt monohydrate, glycine, n,n-bis carboxymethyl-, trisodium salt, monohydrate, nitrilotriacetic acid, trisodium salt monohydrate
PubChem CID 29194
IUPAC Name trisodium;2-[bis(carboxylatomethyl)amino]acetate;hydrate
SMILES C(C(=O)[O-])N(CC(=O)[O-])CC(=O)[O-].O.[Na+].[Na+].[Na+]

Specifications

Melting Point >320.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Plastic bottle
Linear Formula N(CH2CO2Na)3·H2O
Quantity 25 g
Solubility Information Solubility in water: 484g/L (25°C)
Formula Weight 275.09
Percent Purity 99+%
Physical Form Powder, Crystals or Chunks
Chemical Name or Material Nitrilotriacetic acid, trisodium salt, monohydrate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Suspected of causing cancer.
Harmful if swallowed.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Use personal protective equipment as required.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present an

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.