Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCR Safety NitriShield™ with Grippaz™ Technology, Orange Nitrile Gloves

Catalog No. 19812748
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

The unique fish-scale pattern of these orange nitrile gloves provides unsurpassed grip and durability. Ergonomically designed for wet grip on the outside and to reduce hand slippage on the inside.

  • 6 mil, Orange Powder Free Nitrile
  • Fish-scale pattern grip
  • Textured inside for better traction
  • 9.5 Inch Industrial/food service grade
  • Component materials comply with all relevant federal regulations for food contact

Specifications

Certifications/Compliance FDA Compliant
Applications Industrial
AQL 1.5
Color Orange
Disposable Single-use
Exterior Finish Fully Textured
Latex Free Yes
Material Nitrile
Powdered Powder Free
Size X-Large
Sterility Non-sterile
Thickness (English) 6 mil
Thickness (Metric) 0.15 mm
Model 6016OXL
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.