Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitrite Standard, 1000 ppm NO2- (304.5 ppm N), Ricca Chemical
SDP

Catalog No. p-2916755
Click to view available options
Quantity:
120 mL
500 mL
Packaging:
Natural Poly Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 7732-18-5

Specifications

Packaging Natural Poly Bottle
Equivalence 1mL = 1mg NO2 (1000ppm NO2),304.5ppm Nitrogen (N)
Boiling Point 100°C
Quantity 120 mL
Chemical Name or Material Nitrite Standard
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.