Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

nitro-α/β-Synuclein Mouse anti-Human, Clone: nSyn12, MilliporeSigma™

Catalog No. 36011I25UL Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
36011I25UL 25 μL
36011I100UL 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 36011I25UL Supplier MilliporeSigma Supplier No. 36011I25UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

nitro-α/β-Synuclein Monoclonal antibody specifically detects nitro-α/β-Synuclein in Human samples. It is validated for Immunohistochemistry (Paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen nitro-α/β-Synuclein
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone nSyn12
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Host Species Mouse
Immunogen Full-length human recombinant Synuclein nitrated in vitro.
Purification Method Protein G purified
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Stable for 1 year at +2°C to +8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.