Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitrobenzene, 99%

Catalog No. p-7080861
Click to view available options
Quantity:
10,000 g
2500 g
500 g

CAS: 98-95-3 | C6H5NO2 | 123.111 g/mol

Nitrobenzene is used as a solvent and as a starting material in the organic synthesis for the preparation of aniline, benzidine, azobenzene, nitrosobenzene, phenylhydroxylamine and acetaminophen. It is also used as a solvent for electrophilic reagents in synthetic chemistry. Further, it is used to prevent the unpleasant odor in shoe, floor and metal polishes. It acts as a precursor to rubber chemicals, azo dyes and pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Nitrobenzene is used as a solvent and as a starting material in the organic synthesis for the preparation of aniline, benzidine, azobenzene, nitrosobenzene, phenylhydroxylamine and acetaminophen. It is also used as a solvent for electrophilic reagents in synthetic chemistry. Further, it is used to prevent the unpleasant odor in shoe, floor and metal polishes. It acts as a precursor to rubber chemicals, azo dyes and pharmaceuticals.

Solubility
Miscible with ethanol, diethyl ether, acetone, benzene and organic solvents. Slightly miscible with water and carbon tetrachloride.

Notes
Incompatible with strong oxidizing agents, strong reducing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-95-3
Molecular Formula C6H5NO2
Molecular Weight (g/mol) 123.111
MDL Number MFCD00007043
InChI Key LQNUZADURLCDLV-UHFFFAOYSA-N
Synonym nitrobenzol, benzene, nitro, oil of mirbane, mirbane oil, essence of mirbane, nitro-benzene, nitrobenzeen, oil of myrbane, nitrobenzen, mononitrobenzene
PubChem CID 7416
ChEBI CHEBI:27798
IUPAC Name nitrobenzene
SMILES C1=CC=C(C=C1)[N+](=O)[O-]

Specifications

Melting Point 5°C to 6°C
Density 1.196
Boiling Point 210°C to 212°C
Flash Point 87°C (188°F)
Refractive Index 1.552
Quantity 500 g
UN Number UN1662
Beilstein 507540
Merck Index 14,6588
Solubility Information Miscible with ethanol,diethyl ether,acetone,benzene and organic solvents. Slightly miscible with water and carbon tetrachloride.
Formula Weight 123.11
Percent Purity 99%
Chemical Name or Material Nitrobenzene
Show More Show Less
Hazard Category H227-H301+H311+H331-H350-H360F-H372
Hazard Statement GHS H Statement
H300-H310-H330-H372-H351-H361-H227
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing cancer.
Suspected of damaging fertility or the unborn child.
Combustible liquid.
Precautionary Statement P201-P202-P210-P235-P260-P264b-P270-P271-P281-P301+P310-P302+P352-P304+P340-P308+P313-P311-P312-P330-P361-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: NITROBENZENE
EINECSNumber 202-716-0
RTECSNumber DA6475000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.