Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Nitrobenzofurazan Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA185035
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA185035 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA185035 Supplier Invitrogen™ Supplier No. PA185035
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Rabbit anti 4-Fluoro-7-Nitrobenzofurazan antibody recognizes 4-fluoro-7-nitrobenzofurazan (NBD), a fluorescent compund useful for the labeling of amino acids and other biomolecules and also a commonly used derivatizing agent for HPLC.

4-Chloro-7-nitrobenzofurazan is a benzofuran derivative used as a protein reagent since the terminal N-NBD-protein conjugate possesses interesting fluorescence and spectral properties. It has also been used as a covalent inhibitor of both beef heart mitochondrial ATPase and bacterial ATPase. [National Center for Biotechnology Information. PubChem Compound Database; CID=25043].
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nitrobenzofurazan
Applications ELISA
Classification Polyclonal
Concentration 5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene Alias 3-benzoxadiazole; 4-Chloro-7-nitro-2; 4-Chloro-7-nitrobenzo-2-oxa-1; 4-Fluoro-7-Nitrobenzofurazan; 7-Chloro-4-nitrobenzofurazan; C6H2ClN3O3; Nbd chloride
Host Species Rabbit
Immunogen NBD-F conjugated to cationised BSA.
Purification Method Protein G
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Target Molecule Chemical
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.