Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Azure Biosystems Nitrocellulose Transfer Membrane
SDP

Catalog No. AC2106AZ Shop All Azure Biosystems Products
Change view
Click to view available options
Pore Size:
0.22 μm
0.45 μm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Pore Size
AC2106AZ 0.45 μm
AC2107AZ 0.22 μm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC2106AZ Supplier Azure Biosystems Supplier No. AC2106
Add to Cart
Add to Cart

Pre-cut membrane, 8 x 10 cm, mini vertical gels, 10 sheets

  • Pre-cut Nitrocellulose membrane for Western blotting
  • Low-fluorescence Nitrocellulose membranes are available in both 0.45 μm and 0.22 μm pore sizes to accommodate larger and smaller proteins, respectively. We recommend using the .45 μm pore size for the most analytical blotting: protein, ssDNA, and RNA transfers.
  • Low molecular weight proteins
  • Typically used for chemiluminescence
  • Not ideal for stripping
  • Excellent binding properties
  • Quantity: 10 membranes each

Specifications

Length (Metric) 8 cm
Width (Metric) 10 cm
For Use With (Application) Western Blot
Format Sheet
Material 100% Pure Nitrocellulose
Pore Size 0.45 μm
Dimensions (LxW) 8 x 10 cm
Sufficient For 10 Blots
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.