Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitrofurazone 98.0+%, TCI America™
SDP

Catalog No. N0200250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N0200250G 250 g
N020025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N0200250G Supplier TCI America Supplier No. N0200250G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 59-87-0
Molecular Formula C6H6N4O4
Molecular Weight (g/mol) 198.14
MDL Number MFCD00003225
InChI Key IAIWVQXQOWNYOU-FPYGCLRLSA-N
Synonym nitrofurazone, nitrofural, furacilin, furacin, furacillin, aldomycin, furacine, furaldon, nifuzon, actin-n
PubChem CID 5447130
IUPAC Name [(E)-[(5-nitrofuran-2-yl)methylidene]amino]urea
SMILES NC(=O)N\N=C\C1=CC=C(O1)[N+]([O-])=O

Specifications

Melting Point 244°C
Color Yellow
pH 6.0 to 6.5 (saturated H2O)
Quantity 250 g
Formula Weight 198.14
Percent Purity ≥98.0% (HPLC,E)
Physical Form Crystalline Powder
Chemical Name or Material Nitrofurazone
RTECSNumber LT7700000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.