Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nitrate Nitrogen Standard, 100 ppm N (442.7 ppm NO3-), Ricca Chemical
Shop All Ricca Chemical Company Products

Click to view available options
Quantity:
120 mL
500 mL
1 L
Packaging:
Natural Poly Bottle
Specifications
Specifications
| Packaging | Natural Poly Bottle |
| CAS Min % | 98.0, 0.07 |
| CAS Max % | 100.0, 0.07 |
| Boiling Point | 100°C |
| Quantity | 120 mL |
| Chemical Name or Material | Nitrogen Standard, 1 mL = 1 mg N as NO3 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction